Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

470430

Sigma-Aldrich

2-(Trimethylsilyl)phenyl trifluoromethanesulfonate

97%

Synonym(s):

2-(Trimethylsilyl)phenyl 2,2,2-trifluoroacetate, 2-(Trimethylsilyl)phenyl triflate, 2-(Trimethylsilyl)phenyl trifluormethanesulfonate, 2-(Trimethylsilyl)phenyl trifluoromethansulfonate, Trifluoromethanesulfonic acid 2-(trimethylsilyl)phenyl ester, o-Trimethylsilylphenyl triflate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3SO3C6H4Si(CH3)3
CAS Number:
Molecular Weight:
298.35
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.456 (lit.)

bp

70 °C/2 mmHg (lit.)

density

1.229 g/mL at 25 °C (lit.)

functional group

fluoro
triflate

SMILES string

C[Si](C)(C)c1ccccc1OS(=O)(=O)C(F)(F)F

InChI

1S/C10H13F3O3SSi/c1-18(2,3)9-7-5-4-6-8(9)16-17(14,15)10(11,12)13/h4-7H,1-3H3

InChI key

XBHPFCIWRHJDCP-UHFFFAOYSA-N

General description

2-(Trimethylsilyl)phenyl trifluoromethanesulfonate is an important ortho-benzyne precursor in aryne chemistry.

Application

2-(Trimethylsilyl)phenyl trifluoromethanesulfonate may be used to generate benzyne under mild conditions of simple fluoride treatment at room temperature.

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Catherine Hall et al.
Chemical communications (Cambridge, England), 49(69), 7602-7604 (2013-07-24)
A novel transformation of 2-(trimethylsilyl)phenyl trifluoromethanesulfonate aryne precursors is described.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service