Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

470201

Sigma-Aldrich

2-(Propylamino)ethanol

≥98%

Synonym(s):

N-Propylethanolamine

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
$151.00

$151.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 ML
$151.00

About This Item

Linear Formula:
CH3CH2CH2NHCH2CH2OH
CAS Number:
Molecular Weight:
103.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$151.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98%

refractive index

n20/D 1.4415 (lit.)

bp

182 °C (lit.)

density

0.9 g/mL at 25 °C (lit.)

SMILES string

CCCNCCO

InChI

1S/C5H13NO/c1-2-3-6-4-5-7/h6-7H,2-5H2,1H3

InChI key

BCLSJHWBDUYDTR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

172.4 °F - closed cup

Flash Point(C)

78 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Xue Liu et al.
Polymers, 12(11) (2020-10-30)
Thermoresponsive poly(2-(N-alkylacrylamide) ethyl acetate)s with different N-alkyl groups, including poly(2-(N-methylacrylamide) ethyl acetate) (PNMAAEA), poly(2-(N-ethylacrylamide) ethyl acetate) (PNEAAEA), and poly(2-(N-propylacrylamide) ethyl acetate) (PNPAAEA), as well as poly(N-acetoxylethylacrylamide) (PNAEAA), were synthesized by solution RAFT polymerization. Unexpectedly, it was found that there are
CO2 solubility and species distribution in aqueous solutions of 2-(isopropylamino) ethanol and its structural isomers.
Yamada H, et al.
International Journal of Greenhouse Gas Control, 17, 99-105 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service