Skip to Content
MilliporeSigma
All Photos(1)

Documents

469904

Sigma-Aldrich

Tri(propylene glycol) propyl ether, mixture of isomers

97%

Synonym(s):

Tripropylene glycol n -propyl ether, Tripropylene glycol monopropyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2O(C3H6O)2C3H6OH
CAS Number:
Molecular Weight:
234.33
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

refractive index

n20/D 1.43 (lit.)

surface tension

29.8 dyn/cm, 25 °C

viscosity

6.3 cP(25 °C)(lit.)

bp

261 °C (lit.)

solubility

H2O: soluble

density

0.935 g/mL at 25 °C (lit.)

SMILES string

CCCOCC(C)OCC(C)OCC(C)O

InChI

1S/C12H26O4/c1-5-6-14-8-11(3)16-9-12(4)15-7-10(2)13/h10-13H,5-9H2,1-4H3

InChI key

JKEHLQXXZMANPK-UHFFFAOYSA-N

Application

Cosolvent in water-based coatings and cleaners.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service