Skip to Content
MilliporeSigma
All Photos(1)

Documents

469866

Sigma-Aldrich

Tetrakis(diethylamido)titanium(IV)

99.999% trace metals basis

Synonym(s):

TDEAT, Tetrakis(diethylamino)titanium(IV), Titanium(IV) tetrakis(diethylamide)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(C2H5)2N]4Ti
CAS Number:
Molecular Weight:
336.38
Beilstein:
3621431
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

99.999% trace metals basis

form

liquid

reaction suitability

core: titanium
reagent type: catalyst

refractive index

n20/D 1.536 (lit.)

bp

112 °C/0.1 mmHg (lit.)

density

0.931 g/mL at 25 °C (lit.)

SMILES string

CCN(CC)[Ti](N(CC)CC)(N(CC)CC)N(CC)CC

InChI

1S/4C4H10N.Ti/c4*1-3-5-4-2;/h4*3-4H2,1-2H3;/q4*-1;+4

InChI key

VJDVOZLYDLHLSM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Please inquire for bulk quantity, pricing, and packaging options.

Application

Precursor to titanium nitride (TiN) thin films by organometallic chemical vapor deposition (OMCVD).

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Corr. 1B - Water-react 1

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

-19.8 °F - closed cup

Flash Point(C)

-28.8 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thin Solid Films, 85, 2471-2471 (1994)

Articles

Titanium dioxide applications: Semiconducting material characteristics and diverse functionalities.

Nanomaterials are considered a route to the innovations required for large-scale implementation of renewable energy technologies in society to make our life sustainable.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service