Skip to Content
MilliporeSigma
All Photos(2)

Documents

469572

Sigma-Aldrich

Boc-GABA-OH

97%, for peptide synthesis

Synonym(s):

γ-(Boc-amino)butyric acid, 4-(tert-Butoxycarbonylamino)butyric acid, Boc-GABA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3COCONH(CH2)3CO2H
CAS Number:
Molecular Weight:
203.24
Beilstein:
1869578
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

product name

Boc-GABA-OH, 97%

Assay

97%

reaction suitability

reaction type: Boc solid-phase peptide synthesis

bp

78-82 °C/0.2 mmHg (lit.)

mp

55-57 °C (lit.)

application(s)

peptide synthesis

SMILES string

CC(C)(C)OC(=O)NCCCC(O)=O

InChI

1S/C9H17NO4/c1-9(2,3)14-8(13)10-6-4-5-7(11)12/h4-6H2,1-3H3,(H,10,13)(H,11,12)

InChI key

HIDJWBGOQFTDLU-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service