Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

469564

Sigma-Aldrich

cis-1,8-Diamino-p-menthane

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H22N2
CAS Number:
Molecular Weight:
170.30
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

refractive index

n20/D 1.48 (lit.)

bp

233 °C (lit.)

density

0.918 g/mL at 25 °C (lit.)

SMILES string

CC(C)(N)[C@H]1CC[C@@](C)(N)CC1

InChI

1S/C10H22N2/c1-9(2,11)8-4-6-10(3,12)7-5-8/h8H,4-7,11-12H2,1-3H3/t8-,10+

InChI key

KOGSPLLRMRSADR-WAAGHKOSSA-N

General description

cis-1,8-Diamino-p-menthane may be used in the synthesis of cis-(N,N′-p-menth-1,8-ylene)bis(2-bromo-3,3-dimethylbutanamide).

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

THE SYNTHESIS, PHYSICAL, AND SPECTRAL PROPERTIES, AND SOME REACTIONS OF A NEW STABLE bis-a-LACTAM (AZIRIDINONE) WITH A TERPENE SKELETON.
Lengyel I, et al.
Synthetic Communications, 31(23), 3671-3683 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service