All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C6H3ClF3N
CAS Number:
Molecular Weight:
181.54
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
form
solid
mp
36-40 °C (lit.)
SMILES string
FC(F)(F)c1cccnc1Cl
InChI
1S/C6H3ClF3N/c7-5-4(6(8,9)10)2-1-3-11-5/h1-3H
InChI key
RXATZPCCMYMPME-UHFFFAOYSA-N
General description
2-Chloro-3-(trifluoromethyl)pyridine is a trifluoromethyl-substituted pyridine derivative.
Application
2-Chloro-3-(trifluoromethyl)pyridine may be used in the preparation of 5,5′-bis(trifluoromethyl)-2,2′-bipyridine, via modified Ullmann reaction.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B - STOT RE 1
Storage Class Code
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Modified Ullmann coupling of 2-chloro-3-trifluoromethylpyridine.
Munavalli S, et al.
Journal of Fluorine Chemistry, 73(1), 1-6 (1995)
Trifluoromethyl-Substituted Pyridines Through Displacement of Iodine by in situ Generated (Trifluoromethyl) copper.
Cottet F and Schlosser M.
European Journal of Organic Chemistry, 2, 327-330 (2002)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service