Skip to Content
MilliporeSigma
All Photos(1)

Documents

469238

Sigma-Aldrich

4-Fluorophenyl acetate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2C6H4F
CAS Number:
Molecular Weight:
154.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

refractive index

n20/D 1.48 (lit.)

bp

197 °C (lit.)

density

1.178 g/mL at 25 °C (lit.)

SMILES string

CC(=O)Oc1ccc(F)cc1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service