Skip to Content
MilliporeSigma
All Photos(3)

Documents

469106

Sigma-Aldrich

Boc-Tyr-OMe

97%

Synonym(s):

N-(tert-Butoxycarbonyl)-L-tyrosine methyl ester, Boc-L-tyrosine methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(HO)C6H4CH2CH[NHCO2C(CH3)3]CO2HCH3
CAS Number:
Molecular Weight:
295.33
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

optical activity

[α]22/D +51°, c = 1 in chloroform

reaction suitability

reaction type: Boc solid-phase peptide synthesis

mp

100-104 °C (lit.)

application(s)

peptide synthesis

SMILES string

COC(=O)[C@H](Cc1ccc(O)cc1)NC(=O)OC(C)(C)C

InChI

1S/C15H21NO5/c1-15(2,3)21-14(19)16-12(13(18)20-4)9-10-5-7-11(17)8-6-10/h5-8,12,17H,9H2,1-4H3,(H,16,19)/t12-/m0/s1

InChI key

NQIFXJSLCUJHBB-LBPRGKRZSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jie Zeng et al.
Macromolecular bioscience, 16(3), 363-370 (2015-11-28)
The storage method for living cells is one of the major challenges in cell-based applications. Here, a novel supramolecular gel cryopreservation system (BDTC gel system) is introduced, which can observably increase the neural cell viability during cryopreservation process because this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service