Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

469041

Sigma-Aldrich

Hafnium(IV) trifluoromethanesulfonate hydrate

Synonym(s):

Hafnium(IV) triflate, Hf(OTf)4

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3SO3)4Hf
CAS Number:
Molecular Weight:
774.77 (anhydrous basis)
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

core: hafnium
reagent type: catalyst

mp

>350 °C (lit.)

SMILES string

FC(F)(F)S(=O)(=O)O[Hf](OS(=O)(=O)C(F)(F)F)(OS(=O)(=O)C(F)(F)F)OS(=O)(=O)C(F)(F)F

InChI

1S/4CHF3O3S.Hf/c4*2-1(3,4)8(5,6)7;/h4*(H,5,6,7);/q;;;;+4/p-4

InChI key

BQYMOILRPDTPPJ-UHFFFAOYSA-J

Application

Catalyst for:
  • Homogeneous methoxycarbonylation and hydrocarboxylation reactions of phenylacetylene
  • Direct Friedel-Crafts reactions of chromene hemiacetals
  • Aminomethylation reactions under Lewis acidic conditions
  • Prins-type cyclization reactions
  • Direct polycondensation of lactic acid
  • Cationic benzylation reactions
  • Chemoselective thioacetalization and transthioacetalization of carbonyl compounds
Excellent and recyclable catalyst for the mononitration of o-nitrotoluene.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hafnium trifluoromethanesulfonate (Hf (OTf)4) as an efficient catalyst in the fries rearrangement and direct acylation of phenol and naphthol derivatives.
Kobayashi S, et al.
Tetrahedron Letters, 37(12), 2053-2056 (1996)
Waller, F.J. et al.
Tetrahedron Letters, 39, 1641-1641 (1998)
Norio Sakai et al.
The Journal of organic chemistry, 68(2), 483-488 (2003-01-18)
The copper(II) triflate- and hafnium(IV) triflate-catalyzed aminomethylation of indole (2) with an N-silyl-N,O-acetal 1 containing a trichloromethyl group provides the primary amine derivative (3a) in modest yield. When 1 equiv of trimethylchlorosilane (TMSCl) was added to the reaction mixture, the

Articles

Quantum dots (QDs): Semiconductor nanoparticles with diverse applications including displays, lighting, and biomedical imaging.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service