Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

468916

Sigma-Aldrich

[3S-(3α,4aβ,8aβ)]-N-(tert-Butyl)decahydro-3-isoquinolinecarboxamide

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H26N2O
CAS Number:
Molecular Weight:
238.37
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

optical activity

[α]22/D −71°, c = 1 in methanol

mp

112-115 °C (lit.)

SMILES string

CC(C)(C)NC(=O)[C@@H]1C[C@@H]2CCCC[C@@H]2CN1

InChI

1S/C14H26N2O/c1-14(2,3)16-13(17)12-8-10-6-4-5-7-11(10)9-15-12/h10-12,15H,4-9H2,1-3H3,(H,16,17)/t10-,11+,12-/m0/s1

InChI key

UPZBXVBPICTBDP-TUAOUCFPSA-N

Application

Fragment present in various HIV-1 protease inhibitors currently in clinical trials for AIDS treatment.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Trova, M.P. et al.
Bioorganic & Medicinal Chemistry Letters, 3, 1595-1595 (1993)
T J Tucker et al.
Journal of medicinal chemistry, 35(14), 2525-2533 (1992-07-20)
A series of HIV-1 protease inhibitors containing a novel hydroxyethyl secondary amine transition state isostere has been synthesized. The compounds exhibit a strong preference for the (R) stereochemistry at the transition state hydroxyl group. Molecular modeling studies with the prototype

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service