Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

468428

Sigma-Aldrich

4-Chloro-1,2-difluorobenzene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3F2
CAS Number:
Molecular Weight:
148.54
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

refractive index

n20/D 1.475 (lit.)

bp

126 °C (lit.)

density

1.33 g/mL at 25 °C (lit.)

SMILES string

Fc1ccc(Cl)cc1F

InChI

1S/C6H3ClF2/c7-4-1-2-5(8)6(9)3-4/h1-3H

InChI key

OPQMRQYYRSTBME-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Chloro-1,2-difluorobenzene is a halogenated benzene derivative. Its proton shielding has been computed using ab initio methods.[1]

Application

4-Chloro-1,2-difluorobenzene may be used in the preparation of 1-chloro-4,5-difluoro-2-nitrobenzene.[2]

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

96.0 °F - closed cup

Flash Point(C)

35.56 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fumihito Muro et al.
Bioorganic & medicinal chemistry, 16(23), 9991-10000 (2008-10-28)
Optimization of benzoic acid derivatives by introducing substituents into the diphenyl urea moiety led to the identification of compound 20l as a potent VLA-4 antagonist. Compound 20l inhibited eosinophil infiltration into bronchial alveolar lavage fluid in a murine asthma model
An investigation of descriptors based on the critical points in the electron density by building quantitative structure-property relationships for proton chemical shifts.
Buttingsrud B, et al.
Journal of Molecular Structure: THEOCHEM, 810(1), 15-24 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service