Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

467812

Sigma-Aldrich

Potassium carbonate

ReagentPlus®, 99.99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2CO3
CAS Number:
Molecular Weight:
138.21
Beilstein:
4267587
EC Number:
MDL number:
UNSPSC Code:
12352302
Pricing and availability is not currently available.

product line

ReagentPlus®

Assay

99.99%

impurities

≤0.001% N compounds
≤0.004% S compounds
≤0.005% silica
≤0.01% NH4OH ppt.
≤0.01% insolubles
<200 ppm total metallic impurities

mp

891 °C (lit.)

anion traces

chloride (Cl-): ≤0.003%
phosphate (PO43-): ≤0.001%

cation traces

Ca and Mg precipitate: ≤0.01%
Fe: ≤5 ppm
Na: ≤0.02%
heavy metals: ≤5 ppm

SMILES string

[K+].[K+].[O-]C([O-])=O

InChI

1S/CH2O3.2K/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2

InChI key

BWHMMNNQKKPAPP-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service