Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

466174

Sigma-Aldrich

2H-1,3-Benzoxazine-2,4(3H)-dione

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H5NO3
CAS Number:
Molecular Weight:
163.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

form

solid

mp

228-232 °C (lit.)

SMILES string

O=C1NC(=O)c2ccccc2O1

InChI

1S/C8H5NO3/c10-7-5-3-1-2-4-6(5)12-8(11)9-7/h1-4H,(H,9,10,11)

InChI key

OAYRYNVEFFWSHK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Kinetic studies of the alkaline hydrolysis of 2H-1,3-benzoxazine-2,4(3H)-dione (carsalam) were conducted and reaction was found to be of fractional order with respect to [OH-].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A H Kahns et al.
Acta pharmaceutica Nordica, 3(1), 45-50 (1991-01-01)
The kinetics and mechanism of hydrolysis of 1,3-benzoxazine-2,4-dione and its N-methyl and N-benzoyl derivatives were studied in aqueous solution to provide basic information on the reactivity of the benzoxazinedione structure and to assess the potential of unsubstituted 1,3-benzoxazine-2,4-dione as a
Abdullah S Al-Ayed et al.
Journal of colloid and interface science, 361(1), 205-211 (2011-06-11)
The alkaline hydrolysis of carsalam (2H-1,3-benzoxazine-2,4(3H)-dione), denoted as I, and its N-substituted derivatives i.e., N-methyl-1,3-benzoxazine-2,4-dione (II) and N-benzoyl-1,3-benzoxazine-2,4-dione (III) was studied spectrophotometrically at physiological temperature. The rate of hydrolysis was found to be independent on the substrate concentration. In case
A Kamal
Biochemical pharmacology, 40(7), 1669-1671 (1990-10-01)
The metabolites of 4003/2 observed in vivo have been produced by incubation in vitro of 4003/2 with liver post-mitochondrial supernatants from rat, rabbit and dog. The metabolites were characterized by UV, i.r., NMR, MS and HPLC. All the metabolites detected

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service