Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

465216

Sigma-Aldrich

N-Ethylglycine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5NHCH2CO2H
CAS Number:
Molecular Weight:
103.12
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

98%

mp

186 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

CCNCC(O)=O

InChI

1S/C4H9NO2/c1-2-5-3-4(6)7/h5H,2-3H2,1H3,(H,6,7)

InChI key

YPIGGYHFMKJNKV-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Esther Bailón-García et al.
Materials science & engineering. C, Materials for biological applications, 100, 598-607 (2019-04-06)
In the present study carbon xerogels are used for the first time to study the fibroblast cell growth. For that, carbon xerogel microspheres are synthesized and thereafter functionalized with carbon nanofibers followed by the 1,3-dipolar cycloaddition of azomethine ylides (the
Shawn M Sternisha et al.
Biophysical journal, 118(5), 1109-1118 (2020-02-06)
Human glucokinase (GCK) is the prototypic example of an emerging class of proteins with allosteric-like behavior that originates from intrinsic polypeptide dynamics. High-resolution NMR investigations of GCK have elucidated millisecond-timescale dynamics underlying allostery. In contrast, faster motions have remained underexplored
Naoko Arashida et al.
Analytica chimica acta, 954, 77-87 (2017-01-14)
Amino acids and their related metabolites play important roles in various physiological processes and have consequently become biomarkers for diseases. However, accurate quantification methods have only been established for major compounds, such as amino acids and a limited number of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service