Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

464767

Sigma-Aldrich

Tetraammineplatinum(II) tetrachloroplatinate(II)

97%

Synonym(s):

Magnus Green salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[Pt(NH3)4](PtCl4)
CAS Number:
Molecular Weight:
600.09
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

Assay

97%

mp

320 °C (dec.) (lit.)

density

4 g/mL at 25 °C (lit.)

SMILES string

N[Pt++](N)(N)N.Cl[Pt--](Cl)(Cl)Cl

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jovita R Tauro et al.
Journal of biomaterials science. Polymer edition, 16(10), 1233-1244 (2005-11-05)
The objective of this study was to synthesize and characterize particles as a drug-delivery platform for gliomas, a highly advanced and invasive stage of brain tumor with poor prognosis. Poly(aminoethyl methacrylate-co-methyl methacrylate) particles were prepared by suspension polymerization and poly(aminoethyl
B C Chu et al.
Nucleic acids research, 18(17), 5163-5171 (1990-09-11)
We have investigated the efficiency with which PtII complexes cross-link phosphorothioates of oligonucleotides to complementary DNA targets. The A and G residues 2-5 bases downstream from the 5'-phosphorothioate group are preferred sites for cross-linking. Replacement of residues in this part
H C Schuppe et al.
International archives of allergy and immunology, 112(2), 125-132 (1997-02-01)
The sensitizing properties of different complex salts of platinum were defined in vivo by means of the popliteal lymph node (PLN) assay in mice. Hexa- and tetrachloroplatinates were confirmed to be highly immunogenic, inducing vigorous primary immune responses in the
E Thanasias et al.
Advances in experimental medicine and biology, 755, 47-53 (2012-07-25)
Allergic immediate-type reactions by halogenated compounds of platinum (Pt) (platinum salts) have been described in workers in precious metal refineries and catalyst productions. In both industries there are exposures to many different Pt compounds. It is believed that the most
M Becker et al.
Journal of magnetic resonance (San Diego, Calif. : 1997), 133(1), 115-122 (1998-07-09)
The anticancer drug carboplatin has been monitored in rats during treatment by means of in vivo 195Pt NMR spectroscopy at 2.0 T. The purpose of the study was to assess local disposition kinetics in intact tissue following subcutaneous injection of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service