Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

464546

Sigma-Aldrich

Boc-4-phenyl-Phe-OH

97%

Synonym(s):

(S)-2-(Boc-amino)-3-(4-biphenylyl)propionic acid, Boc-4-phenyl-L-phenylalanine, Boc-Bip-OH

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H4CH2CH[NHCO2C(CH3)3]CO2H
CAS Number:
Molecular Weight:
341.40
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

optical activity

[α]22/D +21°, c = 2 in methanol

reaction suitability

reaction type: Boc solid-phase peptide synthesis

mp

103-107 °C (lit.)

application(s)

peptide synthesis

SMILES string

CC(C)(C)OC(=O)N[C@@H](Cc1ccc(cc1)-c2ccccc2)C(O)=O

InChI

1S/C20H23NO4/c1-20(2,3)25-19(24)21-17(18(22)23)13-14-9-11-16(12-10-14)15-7-5-4-6-8-15/h4-12,17H,13H2,1-3H3,(H,21,24)(H,22,23)/t17-/m0/s1

InChI key

NBVVKAUSAGHTSU-KRWDZBQOSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service