Skip to Content
MilliporeSigma
All Photos(2)

Documents

464139

Sigma-Aldrich

6-Methoxysalicylic acid

98%

Synonym(s):

2-Hydroxy-6-methoxybenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H3(OH)CO2H
CAS Number:
Molecular Weight:
168.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

134-138 °C (lit.)

SMILES string

COc1cccc(O)c1C(O)=O

InChI

1S/C8H8O4/c1-12-6-4-2-3-5(9)7(6)8(10)11/h2-4,9H,1H3,(H,10,11)

InChI key

AAUQLHHARJUJEH-UHFFFAOYSA-N

General description

6-Methoxysalicylic acid has been reported to exhibit significant analgesic effects.

Application

6-Methoxysalicylic acid has been used to fabricate a dummy template molecule required for the preparation of dummy molecularly imprinted polymers (DMIPs). These polymers are useful for the simultaneous selective removal and enrichment of ginkgolic acids (GAs) during the processing of Ginkgo biloba leaves. It may be employed as internal standard for the determination of acetylsalicylic acid (aspirin, ASA) and its major metabolite, salicylic acid (SA), in animal plasma by LC/MS/MS.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francisco Palacios-Espinosa et al.
Journal of ethnopharmacology, 118(3), 448-454 (2008-06-28)
Brickellia veronicifolia (Kunth) Gray (Asteraceae) (BV) is broadly commercialized for treating gastrointestinal diseases (stomach aches, biliary colics and dyspepsia), arthritis, diabetes and painful inflammatory complaints. In order to complete the preclinical pharmacological profile of BV, first the antinociceptive effect of
Xiangrong Xu et al.
Biomedical chromatography : BMC, 23(9), 973-979 (2009-04-10)
A simple and sensitive analytical method using liquid chromatography-tandem mass spectrometry (LC/MS/MS) for determination of acetylsalicylic acid (aspirin, ASA) and its major metabolite, salicylic acid (SA), in animal plasma has been developed and validated. Both ASA and SA in plasma
Ecem Evrim Çelik et al.
Food research international (Ottawa, Ont.), 108, 286-294 (2018-05-08)
The interactions of coffee and bread crust melanoidins with hydroxycinnamic and hydroxybenzoic acids (HCA/HBA) containing different numbers of -OH and -OCH₃ groups localized at different positions on the aromatic ring were investigated. By doing so, mechanism of the interactions was
Wenhua Ji et al.
Journal of chromatography. A, 1368, 44-51 (2014-12-03)
Dummy molecularly imprinted polymers (DMIPs) for simultaneously selective removal and enrichment of ginkgolic acids (GAs) during the processing of Ginkgo biloba leaves have been prepared. Two dummy template molecule with similar structural skeleton to GAs, 6-methoxysalicylic acid (MOSA, DT-1) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service