Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

463965

Sigma-Aldrich

(3aR-cis)-(+)-3,3a,8,8a-Tetrahydro-2H-indeno[1,2-d]oxazol-2-one

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9NO2
CAS Number:
Molecular Weight:
175.18
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Assay

97%

optical activity

[α]22/D +76°, c = 0.65 in ethyl acetate

optical purity

ee: 99% (HPLC)

mp

203-209 °C (lit.)

SMILES string

O=C1N[C@H]2[C@H](Cc3ccccc23)O1

InChI

1S/C10H9NO2/c12-10-11-9-7-4-2-1-3-6(7)5-8(9)13-10/h1-4,8-9H,5H2,(H,11,12)/t8-,9+/m0/s1

InChI key

XWZLNPUWNUTPAU-DTWKUNHWSA-N

Application

Versatile chiral auxiliary used in a variety of highly diastereoselective reactions such as Michael additions, Diels-Alder reactions, cyclopropanations, and allylations.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Davies, I.W. et al.
Tetrahedron Letters, 36, 7619-7619 (1995)
Akiba, T. et al.
Tetrahedron, 50, 3905-3905 (1994)
Arun K. Ghosh et al.
The Journal of organic chemistry, 61(18), 6175-6182 (1996-09-06)
Sinefungin (1) a nucleoside antibiotic isolated from Streptomyces has been synthesized from D-ribose. Both the C-6 and C-9 stereogenic centers were constructed by efficient asymmetric syntheses. The C-6 amine stereochemistry was set by a highly diastereoselective allylation (>99% de) of
Davies, S.G. Sanganee, H.J.
Tetrahedron Asymmetry, 6, 671-671 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service