Skip to Content
MilliporeSigma
All Photos(1)

Documents

463434

Sigma-Aldrich

2-Amino-N-cyclohexyl-N-methylbenzylamine

98%

Synonym(s):

N-(2-Aminobenzyl)-N-methylcyclohexanamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H4CH2N(C6H11)CH3
CAS Number:
Molecular Weight:
218.34
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

44-47 °C (lit.)

SMILES string

CN(Cc1ccccc1N)C2CCCCC2

InChI

1S/C14H22N2/c1-16(13-8-3-2-4-9-13)11-12-7-5-6-10-14(12)15/h5-7,10,13H,2-4,8-9,11,15H2,1H3

InChI key

MTVBBLNLYSYKCQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rodriguez VG, et al.
Journal of High Resolution Chromatography, 19, 703-703 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service