Skip to Content
MilliporeSigma
All Photos(1)

Documents

462756

Sigma-Aldrich

(+)-N-Benzoylglutamic acid

synthetic, 97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCH2CH2CH(NHCOC6H5)CO2H
CAS Number:
Molecular Weight:
251.24
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:

Assay

97%

optical activity

[α]22/D +12°, c = 5 in H2O

reaction suitability

reaction type: solution phase peptide synthesis

mp

130-135 °C (lit.)

application(s)

peptide synthesis

SMILES string

OC(=O)CC[C@@H](NC(=O)c1ccccc1)C(O)=O

InChI

1S/C12H13NO5/c14-10(15)7-6-9(12(17)18)13-11(16)8-4-2-1-3-5-8/h1-5,9H,6-7H2,(H,13,16)(H,14,15)(H,17,18)/t9-/m1/s1

InChI key

LPJXPACOXRZCCP-SECBINFHSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Webber Wei-Po Lai et al.
Water research, 112, 157-166 (2017-02-06)
Bicarbonate anion (HCO
Ming-Hao Hsu et al.
Journal of hazardous materials, 373, 468-475 (2019-04-03)
Methotrexate, a chemotherapeutic agent, was found to undergo self-sensitized photodegradation in aqueous environments. As the initial concentration increased, methotrexate was able to enhance its own direct photolysis reaction not only in DI but also in natural waters. The methotrexate degradation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service