Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

462675

Sigma-Aldrich

Tin(II) methanesulfonate solution

50 wt. % in H2O

Synonym(s):

Stannous methanesulfonate, Stannous methylsulfonate

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
$91.30

$91.30


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 ML
$91.30

About This Item

Linear Formula:
(CH3SO3)2Sn
CAS Number:
Molecular Weight:
308.91
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$91.30


Please contact Customer Service for Availability

Request a Bulk Order

reaction suitability

core: tin
reagent type: catalyst

Quality Level

concentration

50 wt. % in H2O

refractive index

n20/D 1.444

mp

−27 °C

density

1.55 g/mL at 25 °C

SMILES string

[SnH2++].CS([O-])(=O)=O.CS([O-])(=O)=O

InChI

1S/2CH4O3S.Sn/c2*1-5(2,3)4;/h2*1H3,(H,2,3,4);/q;;+2/p-2

InChI key

JALQQBGHJJURDQ-UHFFFAOYSA-L

Other Notes

Contains methanesulfonic acid

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service