Skip to Content
MilliporeSigma
All Photos(2)

Documents

461784

Sigma-Aldrich

Benzyl (R)-(+)-2-hydroxy-3-phenylpropionate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH(OH)CO2CH2C6H5
CAS Number:
Molecular Weight:
256.30
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

refractive index

n20/D 1.559 (lit.)

bp

228 °C (lit.)

mp

20-30 °C (lit.)

density

1.14 g/mL at 25 °C (lit.)

SMILES string

O[C@H](Cc1ccccc1)C(=O)OCc2ccccc2

InChI

1S/C16H16O3/c17-15(11-13-7-3-1-4-8-13)16(18)19-12-14-9-5-2-6-10-14/h1-10,15,17H,11-12H2/t15-/m1/s1

InChI key

XFULYMQQCZRWQB-OAHLLOKOSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service