Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

461458

Sigma-Aldrich

(R)-(+)-N-(1-Phenylethyl)phthalamic acid

97%

Synonym(s):

(R)-(+)-N-(α-Methylbenzyl)phthalamic acid, (R)-(+)-N-(α-Methylbenzyl)phthalic acid monoamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-[C6H5CH(CH3)NHCO]C6H4CO2H
CAS Number:
Molecular Weight:
269.30
Beilstein:
5793588
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

optical activity

[α]24/D +46°, c = 2 in ethanol

mp

130-135 °C (lit.)

SMILES string

C[C@@H](NC(=O)c1ccccc1C(O)=O)c2ccccc2

InChI

1S/C16H15NO3/c1-11(12-7-3-2-4-8-12)17-15(18)13-9-5-6-10-14(13)16(19)20/h2-11H,1H3,(H,17,18)(H,19,20)/t11-/m1/s1

InChI key

VCFKXWGKKDZMPO-LLVKDONJSA-N

Application

Acid for the resolution of amines.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kozma, D. et al.
J. Therm. Anal., 44, 339-339 (1995)
Acs, M. et al.
Chirality, 6, 314-314 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service