Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

461296

Sigma-Aldrich

Acetic acid, alkyl (C9 to C11) esters mixture

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2R, R = C10H21 (primarily)
CAS Number:
MDL number:
UNSPSC Code:
12162002

refractive index

n20/D 1.429 (lit.)

bp

220-250 °C (lit.)

density

0.87 g/mL at 25 °C (lit.)

General description

Contains various branched and linear isomers of C9 to C11 esters

Application

Coalescing solvent

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service