Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

460680

Sigma-Aldrich

Tricyclohexyltin hydride

97%

Synonym(s):

Tricyclohexylstannane, Tricyclohexyltin, Tricyclohexyltin hydride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H11)3SnH
CAS Number:
Molecular Weight:
369.17
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

97%

reaction suitability

core: tin
reagent type: reductant

refractive index

n20/D 1.5409 (lit.)

bp

129 °C/0.05 mmHg (lit.)

SMILES string

C1CCC(CC1)[SnH](C2CCCCC2)C3CCCCC3

InChI

1S/3C6H11.Sn.H/c3*1-2-4-6-5-3-1;;/h3*1H,2-6H2;;

InChI key

SQEBZYNJTQKFEQ-UHFFFAOYSA-N

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT RE 1 - Water-react 2

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

221.0 °F - closed cup

Flash Point(C)

105 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service