Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

460567

Sigma-Aldrich

Antimony(III) iodide

anhydrous, −10 mesh, 99.999% trace metals basis

Synonym(s):

Antimony triiodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SbI3
CAS Number:
Molecular Weight:
502.47
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

anhydrous

Assay

99.999% trace metals basis

form

liquid

particle size

−10 mesh

bp

401 °C (lit.)

mp

170 °C (lit.)

density

4.92 g/mL at 25 °C (lit.)

SMILES string

I[Sb](I)I

InChI

1S/3HI.Sb/h3*1H;/q;;;+3/p-3

InChI key

KWQLUUQBTAXYCB-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service