Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

460168

Sigma-Aldrich

Poly(ethylene glycol) monooleate

average Mn ~460

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

mol wt

average Mn ~460

refractive index

n20/D 1.465

bp

>260 °C (lit.)

acid number

3 KOH/g

hydroxyl value

90 mg KOH/g

iodine value

3

solubility

toluene, ethanol and acetone: soluble (dispersible in water)

density

0.969 g/mL at 25 °C

SMILES string

OCCO.[H]\C(CCCCCCCC)=C(/[H])CCCCCCCC(O)=O

InChI

1S/C20H38O3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-20(22)23-19-18-21/h9-10,21H,2-8,11-19H2,1H3/b10-9-

InChI key

MUHFRORXWCGZGE-KTKRTIGZSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Plasticizer, surfactant.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service