Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

460133

Sigma-Aldrich

Poly(ethylene glycol) monolaurate

average Mn ~400

Synonym(s):

Polyethylene glycol 400 monolaurate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

mol wt

average Mn ~400

refractive index

n20/D 1.455

bp

>260 °C (lit.)

hydroxyl value

140 mg KOH/g

iodine value

10

solubility

toluene, ethanol and acetone: soluble (dispersible in water)

density

0.985 g/mL at 25 °C

SMILES string

OCCO.CCCCCCCCCCCC(O)=O

InChI

1S/C14H28O3/c1-2-3-4-5-6-7-8-9-10-11-14(16)17-13-12-15/h15H,2-13H2,1H3

InChI key

CTXGTHVAWRBISV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Plasticizer, surfactant.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V V Dmitriev et al.
Mikrobiologiia, 57(2), 223-230 (1988-03-01)
Yeasts capable of utilizing the non-ionogenic surfactant Laurox-9 were identified as Cryptococcus humicolus (Daszewska) Golubev and Rhodotorula mucilaginosa (Jöerg) Harrison. IR spectrometry used for the quantitative assay of Laurox-9 showed that R. mucilaginosa utilized it as a sole carbon source.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service