Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

459933

Sigma-Aldrich

Z-D-Phenylalaninol

97%

Synonym(s):

(R)-(+)-2-(Carbobenzyloxyamino)-3-phenyl-1-propanol, (R)-2-(Z-Amino)-3-phenyl-1-propanol, N-(Carbobenzyloxy)-D-phenylalaninol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH(NHCO2CH2C6H5)CH2OH
CAS Number:
Molecular Weight:
285.34
Beilstein:
2221968
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

optical activity

[α]22/D +30°, c = 1 in chloroform

reaction suitability

reaction type: solution phase peptide synthesis

mp

92-95 °C (lit.)

application(s)

peptide synthesis

SMILES string

OC[C@@H](Cc1ccccc1)NC(=O)OCc2ccccc2

InChI

1S/C17H19NO3/c19-12-16(11-14-7-3-1-4-8-14)18-17(20)21-13-15-9-5-2-6-10-15/h1-10,16,19H,11-13H2,(H,18,20)/t16-/m1/s1

InChI key

WPOFMMJJCPZPAO-MRXNPFEDSA-N

Application

Employed in the synthesis of biochemically active compounds. For references, see Aldrichimica Acta .

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aldrichimica Acta, 28, 13-13 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service