Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

459755

Sigma-Aldrich

Poly(trimellitic anhydride chloride-alt-benzidine) solution

20 wt. % in NMP/xylene

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12162002

dielectric constant

3.8, 1 MHz (50% RH)

concentration

20 wt. % in NMP/xylene

refractive index

n20/D 1.528

bp

177 °C

mp

494 °C (dec.)

transition temp

Tg 300 °C

density

1.07 g/mL at 25 °C

Application

Buffer coating for discrete and integrated circuit devices.

Features and Benefits

Fully imidized polymer.
Self-priming coating with excellent adhesion to a variety of substrates, low ionic content, stable at room temperature.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service