Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

459321

Sigma-Aldrich

Fmoc-Cys(Trt)-OH

97%

Synonym(s):

N-(9-Fluorenylmethoxycarbonyl)-S-trityl-L-cysteine, Nα-Fmoc-S-trityl-L-cysteine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H31NO4S
CAS Number:
Molecular Weight:
585.71
Beilstein:
4221286
MDL number:
UNSPSC Code:
12352103

Assay

97%

optical activity

[α]20/D +24°, c = 1 in methanol

mp

170-173 °C (lit.)

SMILES string

OC(=O)[C@H](CSC(c1ccccc1)(c2ccccc2)c3ccccc3)NC(=O)OCC4c5ccccc5-c6ccccc46

InChI

1S/C37H31NO4S/c39-35(40)34(38-36(41)42-24-33-31-22-12-10-20-29(31)30-21-11-13-23-32(30)33)25-43-37(26-14-4-1-5-15-26,27-16-6-2-7-17-27)28-18-8-3-9-19-28/h1-23,33-34H,24-25H2,(H,38,41)(H,39,40)/t34-/m0/s1

InChI key

KLBPUVPNPAJWHZ-UMSFTDKQSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service