Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

457906

Sigma-Aldrich

Propylene glycol ethyl ether acetate,mixture of isomers

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H14O3
CAS Number:
Molecular Weight:
146.18
MDL number:
UNSPSC Code:
12162002

Assay

95%

refractive index

n20/D 1.404 (lit.)

bp

158 °C (lit.)

density

0.94 g/mL at 25 °C (lit.)

SMILES string

CCOCCCOC(C)=O


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service