Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

457612

Sigma-Aldrich

1-(4-Methoxyphenyl)-1H-imidazole

98%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$69.60

$69.60


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$69.60

About This Item

Empirical Formula (Hill Notation):
C10H10N2O
CAS Number:
Molecular Weight:
174.20
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$69.60


Please contact Customer Service for Availability

Request a Bulk Order

Assay

98%

form

solid

bp

120 °C/0.1 mmHg (lit.)

mp

59-67 °C (lit.)

SMILES string

COc1ccc(cc1)-n2ccnc2

InChI

1S/C10H10N2O/c1-13-10-4-2-9(3-5-10)12-7-6-11-8-12/h2-8H,1H3

InChI key

XNLOIFUGGCCEQX-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enhanced catalytic performance by copper nanoparticle-graphene based composite.
Mondal P, et al.
Royal Society of Chemistry Advances, 3(16), 5615-5623 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service