Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

457477

Sigma-Aldrich

1-Ethynyl-4-pentylbenzene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4C6H4C≡CH
CAS Number:
Molecular Weight:
172.27
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.523 (lit.)

bp

172 °C (lit.)

density

0.885 g/mL at 25 °C (lit.)

SMILES string

CCCCCc1ccc(cc1)C#C

InChI

1S/C13H16/c1-3-5-6-7-13-10-8-12(4-2)9-11-13/h2,8-11H,3,5-7H2,1H3

InChI key

APGNXGIUUTWIRE-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

217.4 °F - closed cup

Flash Point(C)

103.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Berta Gómez-Lor et al.
Chemical communications (Cambridge, England), (48)(48), 5012-5014 (2006-12-06)
Discotic liquid crystals based on triindole, a novel redox active central core, have been synthesized and their mesomorphic behaviour investigated.
Synthesis and Study of the Thermal and Chiro-Optical Properties of Polyacetylenes with Bulky Side Groups: Poly (1-ethynyl-4-biphenyl), Poly (1-ethynyl-4-phenoxybenzene) and Poly (1-ethynyl-4-pentylbenzene).
Cataldo F, et al.
Journal of Macromolecular Science, Part A: Pure and Applied Chemistry, 46(9), 860-869 (2009)
Stable solution-processed high-mobility substituted pentacene semiconductors
Li, Y., Wu, Y., Liu, P., Prostran, Z., Gardner, S., & Ong, B. S.
Chemistry of Materials, 19(3), 418-423 (2007)
Srinivasarao Meneni et al.
Bioorganic & medicinal chemistry, 15(8), 3082-3088 (2007-03-06)
Starting with 5-iodo-2'-deoxyuridine, a series of 5-alkynyl-2'-deoxyuridines (with n-propyl, cyclopropyl, 1-hydroxycyclohexyl, p-tolyl, p-tert-butylphenyl, p-pentylphenyl, and trimethylsilyl alkyne substituents) have been synthesized via the palladium-catalyzed (Sonogashira) coupling reaction followed by a simplified isolation protocol (76-94% yield). The cytotoxic activity of modified

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service