Skip to Content
MilliporeSigma
All Photos(1)

Documents

457299

Sigma-Aldrich

Poly(tetrafluoroethylene oxide-co-difluoromethylene oxide) α,ω-diol, ethoxylated

average Mn ~2,200

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO(CH2CH2O)xCH2CF2O(CF2CF2O)y(CF2O)ZCF2CH2(OCH2CH2)xOH
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

specific activity

~3 ethyl ether units/molecule

mol wt

average Mn ~2,200

refractive index

n20/D 1.311 (lit.)

viscosity

125 cSt(20 °C)(lit.)

density

1.74 g/mL at 25 °C (lit.)

SMILES string

OCCO.OC(O)(F)F.OCC(O)(F)F.OC(F)(F)C(O)(F)F

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service