Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

457280

Sigma-Aldrich

Poly(tetrafluoroethylene oxide-co-difluoromethylene oxide) α,ω-bis(methyl carboxylate)

average Mn ~2,000

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3O2CCF2O(CF2CF2O)x(CF2O)yCF2CO2CH3
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
Pricing and availability is not currently available.

mol wt

average Mn ~2,000

dielectric constant

3.7, 50 Hz (23 °C)

refractive index

n20/D 1.298 (lit.)

surface tension

225 dyn/cm, 20 °C

viscosity

20 cSt(20 °C)(lit.)

acid number

5 meq/kg

solubility

isopropanol, MEK, and THF: soluble

density

1.77 g/mL at 25 °C (lit.)

SMILES string

OC(O)(F)F.COC(=O)C(O)(F)F.OC(F)(F)C(O)(F)F

Application

Topical lubricant.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service