Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

456950

Sigma-Aldrich

m-Xylylenediamine/acrylonitrile adduct

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(CH2NR2)2, R = H or CH2CH2CN
CAS Number:
Molecular Weight:
54.07
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
To inquire about this product 456950, please contact your local MilliporeSigma office or dealer. Contact Technical Service

composition

dicyanoethyl adduct, 65 wt. %
monocyanoethyl adduct, 25 wt. %

refractive index

n20/D 1.542 (lit.)

viscosity

120 cP(25 °C)(lit.)

bp

281 °C (lit.)

solubility

water, ethanol, chloroform and benzene.: soluble

density

0.952 g/mL at 25 °C (lit.)

SMILES string

C=CC#N.NCc1cccc(CN)c1

InChI

1S/C8H12N2.C3H3N/c9-5-7-2-1-3-8(4-7)6-10;1-2-3-4/h1-4H,5-6,9-10H2;2H,1H2

InChI key

XMEXUJUMKRUUNG-UHFFFAOYSA-N

Application

Precursor for polyurethanes. Improves chemical resistance and reduces free amines in Mannich-type hardeners for epoxides.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service