Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

456403

Sigma-Aldrich

2,5-Dibromo-3-dodecylthiophene

97%

Synonym(s):

2,5-Dibromo-3-dodecylthiophene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H26Br2S
CAS Number:
Molecular Weight:
410.25
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

refractive index

n20/D 1.53 (lit.)

bp

135 °C/2.5 mmHg (lit.)

density

1.321 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCc1cc(Br)sc1Br

InChI

1S/C16H26Br2S/c1-2-3-4-5-6-7-8-9-10-11-12-14-13-15(17)19-16(14)18/h13H,2-12H2,1H3

InChI key

ZIZGNWAMVWNAJT-UHFFFAOYSA-N

Application

Conducting polymer precursor.

Legal Information

Product of Rieke Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of the American Chemical Society, 117, 233-233 (1995)

Articles

Oligothiophenes are important organic electronic materials which can be produced using synthetic intermediates and Suzuki coupling.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service