Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

456381

Sigma-Aldrich

2,5-Dibromo-3-decylthiophene

97%

Synonym(s):

2,5-Dibromo-3-decylthiophene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H22Br2S
CAS Number:
Molecular Weight:
382.20
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

refractive index

n20/D 1.538 (lit.)

bp

168-170 °C/2 mmHg (lit.)

density

1.372 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCCCCCCCCCc1cc(Br)sc1Br

InChI

1S/C14H22Br2S/c1-2-3-4-5-6-7-8-9-10-12-11-13(15)17-14(12)16/h11H,2-10H2,1H3

InChI key

CQDSRZZKSUNTSB-UHFFFAOYSA-N

Application

Conducting polymer precursor.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of the American Chemical Society, 117, 233-233 (1995)

Articles

Oligothiophenes are important organic electronic materials which can be produced using synthetic intermediates and Suzuki coupling.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service