Skip to Content
MilliporeSigma
All Photos(1)

Documents

456330

Sigma-Aldrich

1,2,4,5-Benzenetetracarboxylic dianhydride-d2

98 atom % D, 98% (CP)

Synonym(s):

Pyromellitic dianhydride-d2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10D2O6
CAS Number:
Molecular Weight:
220.13
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

98 atom % D

Assay

98% (CP)

form

solid

mp

283-286 °C (lit.)

mass shift

M+2

SMILES string

[2H]c1c2C(=O)OC(=O)c2c([2H])c3C(=O)OC(=O)c13

InChI

1S/C10H2O6/c11-7-3-1-4-6(10(14)16-8(4)12)2-5(3)9(13)15-7/h1-2H/i1D,2D

InChI key

ANSXAPJVJOKRDJ-QDNHWIQGSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service