Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

456233

Sigma-Aldrich

L-Aspartic acid dimethyl ester hydrochloride

97%

Synonym(s):

Dimethyl L-aspartate hydrochloride, L-Asp(OMe)OMe·HCl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3O2CCH2CH(NH2)CO2CH3 · HCl
CAS Number:
Molecular Weight:
197.62
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

reaction suitability

reaction type: solution phase peptide synthesis

mp

115-117 °C (lit.)

application(s)

peptide synthesis

SMILES string

Cl.COC(=O)C[C@H](N)C(=O)OC

InChI

1S/C6H11NO4.ClH/c1-10-5(8)3-4(7)6(9)11-2;/h4H,3,7H2,1-2H3;1H/t4-;/m0./s1

InChI key

PNLXWGDXZOYUKB-WCCKRBBISA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Soojin Kim et al.
Pharmaceutics, 12(9) (2020-09-10)
An elevated level of endoplasmic reticulum (ER) stress is considered an aggravating factor for inflammatory bowel disease (IBD). To develop an ER-stress attenuator that is effective against colitis, 4-phenylbutyric acid (4-PBA), a chemical chaperone that alleviates ER stress, was conjugated
Wooseong Kim et al.
Molecular pharmaceutics, 16(9), 4007-4016 (2019-08-07)
We investigated if the therapeutic switching of sofalcone (SFC), a gastroprotective agent, to an anticolitic agent is feasible using colon-targeted drug delivery. SFC can activate the anti-inflammatory nuclear factor (erythroid-derived 2)-like 2 (Nrf2)-hemeoxygenase-1 (HO-1) pathway in human colon epithelial cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service