Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

455520

Sigma-Aldrich

2-Ethoxyphenylboronic acid

≥95%

Synonym(s):

2-(Ethoxy)benzeneboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OC6H4B(OH)2
CAS Number:
Molecular Weight:
165.98
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95%

form

solid

mp

97-102 °C (lit.)

SMILES string

CCOc1ccccc1B(O)O

InChI

1S/C8H11BO3/c1-2-12-8-6-4-3-5-7(8)9(10)11/h3-6,10-11H,2H2,1H3

InChI key

DGFCTCGCMKEILT-UHFFFAOYSA-N

Application

Reactant involved in Suzuki-Miyaura cross-coupling reactions for synthesis of pyridines and pyrimidines, reactions of bromo-N-methylpyrrole, or synthesis of diarylbenzophenones

Reactant involved in:
  • Ipso-amidation with nitriles
  • Studies of the effect of biaryl phosphine ligand structure on C-N and C-C bond formation
  • Oxidative coupling via C-H arylation

Other Notes

Contains varying amounts of anhydride.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service