Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

454877

Sigma-Aldrich

Diisopropyl (S)-(−)-malate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHO2CCH2CH(OH)CO2CH(CH3)2
CAS Number:
Molecular Weight:
218.25
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

optical activity

[α]20/D −13°, neat

refractive index

n20/D 1.43 (lit.)

bp

237 °C (lit.)

density

1.055 g/mL at 25 °C (lit.)

functional group

ester
hydroxyl

SMILES string

CC(C)OC(=O)C[C@H](O)C(=O)OC(C)C

InChI

1S/C10H18O5/c1-6(2)14-9(12)5-8(11)10(13)15-7(3)4/h6-8,11H,5H2,1-4H3/t8-/m0/s1

InChI key

XYVHRFVONMVDGK-QMMMGPOBSA-N

Application

Diisopropyl (S)-(−)-malate can be used as a starting material in the total synthesis of (−)-wikstromol, an α-hydroxylated lactone lignan found in Asian medicinal plants.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enantioselective Synthesis of (−)-Wikstromol Using a New Approach via Malic Acid
Sefkow M
The Journal of Organic Chemistry, 66(7), 2343-2349 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service