Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

454796

Sigma-Aldrich

Benzyloxyacetaldehyde diethyl acetal

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OCH2CH(OC2H5)2
CAS Number:
Molecular Weight:
224.30
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

refractive index

n20/D 1.478 (lit.)

bp

99-100 °C/0.6 mmHg (lit.)

density

0.987 g/mL at 25 °C (lit.)

SMILES string

CCOC(COCc1ccccc1)OCC

InChI

1S/C13H20O3/c1-3-15-13(16-4-2)11-14-10-12-8-6-5-7-9-12/h5-9,13H,3-4,10-11H2,1-2H3

InChI key

VTYTZCJKJNWMGA-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of novel 1, 3-dioxolane nucleoside analogues.
Cai D-M, et al.
Chin. J. Chem., 22(12), 1425-1431 (2004)
(?)-Dioxolane-T ((?)-1-[(2?, 4?)-2-(hydroxymethyl)-4-dioxolanyl] thymine): A new 2', 3'-dideoxynucleoside prototype within vitro activity against HIV.
Norbeck DW, et al.
Tetrahedron Letters, 30(46), 6263-6266 (1989)
Silas P Cook et al.
The Journal of biological chemistry, 277(31), 27782-27792 (2002-05-22)
Serine racemase (SR) is a brain enzyme present in glial cells, where it isomerizes L-serine into D-serine that, in turn, diffuses and coactivates the N-methyl-D-aspartate receptor through the binding to the so-called "glycine site." We have developed a method for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service