Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

454559

Sigma-Aldrich

1,4-Difluoroanthraquinone

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H6F2O2
CAS Number:
Molecular Weight:
244.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Quality Level

Assay

96%

mp

228-232 °C (lit.)

SMILES string

Fc1ccc(F)c2C(=O)c3ccccc3C(=O)c12

InChI

1S/C14H6F2O2/c15-9-5-6-10(16)12-11(9)13(17)7-3-1-2-4-8(7)14(12)18/h1-6H

InChI key

DTNCXQHDOJRTCD-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heterogeneous organocatalysis for the asymmetric desymmetrization of meso-cyclic anhydrides using silica gel-supported bis-cinchona alkaloids.
Kim HS, et al.
Tetrahedron, 60(52), 12051-12051 (2004)
Synthesis of a New Chiral Ligand (QN)2AQN and Its Application to the Catalytic Asymmetric Aminohydroxylation of Methyl Cinnamates.
Si-Kun C, et al.
Chin. J. Chem., 25(8), 982-986 (2005)
Synthesis of fluorine-substituted anthraquinones and aza-anthraquinones.
Khanapure SP, et al.
Journal of Fluorine Chemistry, 68(2), 131-134 (1994)
H-J Lehmler et al.
Tetrahedron, 66(14), 2561-2569 (2010-03-23)
A series of colored hydrocarbon and fluorocarbon tagged 1-fluoro-4-alkylamino-anthraquinones and 1,4-bis-alkylamino-anthraquinone probe molecules were synthesized from a (fluorinated) alkyl amine and 1,4-difluoroanthraquinone to aid in the development of fluorous separation applications. The anthraquinones displayed stacking of the anthraquinone tricycle and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service