Skip to Content
MilliporeSigma
All Photos(1)

Documents

454257

Sigma-Aldrich

2-Bromo-5-nitroaniline

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H3(NO2)NH2
CAS Number:
Molecular Weight:
217.02
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

139-141 °C (lit.)

SMILES string

Nc1cc(ccc1Br)[N+]([O-])=O

InChI

1S/C6H5BrN2O2/c7-5-2-1-4(9(10)11)3-6(5)8/h1-3H,8H2

InChI key

BAAUCXCLMDAZEL-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An intramolecular palladium-catalysed aryl amination reaction to produce benzimidazoles.
Brain CT and Brunton SA.
Tetrahedron Letters, 43(10), 1893-1895 (2002)
Formation and rearrangement of ipso intermediates in aromatic free-radical chlorination reactions.
Everly CR and Traynham JG.
The Journal of Organic Chemistry, 44(11), 1784-1787 (1979)
Olivier Baudoin et al.
Bioorganic & medicinal chemistry, 10(11), 3395-3400 (2002-09-06)
An improvement of the synthesis of biphenyl-carbamate 2a, the most active analogue of rhazinilam 1 so far, was performed using the Pd-catalyzed borylation/Suzuki coupling (BSC) method developed in our laboratories. The preparation of A-ring analogues of 2a bearing electron-withdrawing or
K Sangeetha et al.
Colloids and surfaces. B, Biointerfaces, 173, 249-257 (2018-10-10)
Exploration of rare earth metals for the Surface Enhanced Raman Scattering (SERS) is greatly preferred to identify probe molecules even at nano molar level. Highly stable Rh nanoparticles (NPs) which are ultra-small size have been prepared within 20 min of reaction

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service