Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

454249

Sigma-Aldrich

2-Methylnaphthalene-d10

98 atom % D

Synonym(s):

ß-Methylnaphthalene-d10

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10D7CD3
CAS Number:
Molecular Weight:
152.26
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

98 atom % D

Quality Level

form

solid

bp

241-242 °C (lit.)

mp

34-36 °C (lit.)

density

1.071 g/mL at 25 °C (lit.)

mass shift

M+10

SMILES string

[2H]c1c([2H])c([2H])c2c([2H])c(c([2H])c([2H])c2c1[2H])C([2H])([2H])[2H]

InChI

1S/C11H10/c1-9-6-7-10-4-2-3-5-11(10)8-9/h2-8H,1H3/i1D3,2D,3D,4D,5D,6D,7D,8D

InChI key

QIMMUPPBPVKWKM-UZHHFJDZSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paulina K Piotrowski et al.
Journal of chromatography. A, 1579, 99-105 (2018-10-22)
Development of shale gas resources through the use of hydraulic fracturing has raised a multitude of environmental concerns and motivated research towards the understanding of shale gas systems. Previous research has demonstrated the potential of utilizing hydrocarbon distributions towards the
F Aladar Bencsath et al.
Analytical and bioanalytical chemistry, 407(14), 4079-4090 (2015-03-23)
A headspace solid-phase microextraction gas chromatography-mass spectrometry (SPME GC-MS) method is described, to screen seafood for volatile organic compounds (VOCs) associated with petrochemical taint. VOCs are extracted from the headspace of heated sample homogenates by adsorption onto a SPME fiber

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service