Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

454141

Sigma-Aldrich

Bis(4-fluorophenyl)phenylphosphine oxide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(FC6H4)2P(O)C6H5
CAS Number:
Molecular Weight:
314.27
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

Assay

97%

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling

mp

125-131 °C (lit.)

functional group

phosphine oxide

SMILES string

Fc1ccc(cc1)P(=O)(c2ccccc2)c3ccc(F)cc3

InChI

1S/C18H13F2OP/c19-14-6-10-17(11-7-14)22(21,16-4-2-1-3-5-16)18-12-8-15(20)9-13-18/h1-13H

InChI key

AAYLOGMTTMROGA-UHFFFAOYSA-N

Application

  • Catalyst for hydroformylation reactions
  • Used for preparation of chlorine-tolerant polymers for desalination
  • Crosslinking agent in preparation of polymer electrolyte membranes for fuel cell applications

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service