Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

453870

Sigma-Aldrich

2-Vinylnaphthalene

optical grade, 98%

Synonym(s):

β-Vinylnaphthalene, (2-Naphthyl)ethene, 2-Ethenylnaphthalene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7CH=CH2
CAS Number:
Molecular Weight:
154.21
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

grade

optical grade

Assay

98%

mp

64-68 °C (lit.)

storage temp.

2-8°C

SMILES string

C=Cc1ccc2ccccc2c1

InChI

1S/C12H10/c1-2-10-7-8-11-5-3-4-6-12(11)9-10/h2-9H,1H2

InChI key

KXYAVSFOJVUIHT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used in plastic scintillators, neutron detectors and other demanding device applications.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT SE 2

Target Organs

Eyes

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuliang Yang et al.
Chemistry, an Asian journal, 11(19), 2797-2801 (2016-04-29)
We report an efficient and controllable route to prepare magnetic tubular nanoreactors composed of a mesoporous SiO

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service